Assay Details
| Gene Symbol: ABHD17C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015142862.2 | XP_014998348.2 | 894 | 115 | ||
| GenBank mRNA | JV589453.1 | - | 414 | 115 | ||
| JV624473.1 | - | 431 | 115 | |||
More Information
Set Membership: |
|

ABHD17C
Genomic Map