Assay Details
| Gene Symbol: SLFNL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015136709.2 | XP_014992195.2 | 1521 | 87 | ||
| XM_015136720.2 | XP_014992206.2 | 1521 | 87 | |||
| XM_015136734.2 | XP_014992220.2 | 1521 | 87 | |||
| XM_015136742.2 | XP_014992228.2 | 1449 | 87 | |||
| XM_015136748.2 | XP_014992234.2 | 1456 | 87 | |||
| XM_015136762.2 | XP_014992248.2 | 1411 | 87 | |||
| XM_015136769.2 | XP_014992255.2 | 1346 | 87 | |||
| XM_028835298.1 | XP_028691131.1 | 1383 | 87 | |||
| GenBank mRNA | JV644663.1 | - | 546 | 87 | ||
More Information
Set Membership: |
|

SLFNL1
Genomic Map