Assay Details
| Gene Symbol: ZNF529 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015123768.2 | XP_014979254.2 | 153 | 130 | ||
| XM_015123774.2 | XP_014979260.2 | 153 | 130 | |||
| GenBank mRNA | JV704285.1 | - | 137 | 130 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015123768.2, XM_015123774.2 transcripts.
Set Membership: |
|

ZNF529
Genomic Map