Assay Details
| Gene Symbol: ZNF529 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015123766.2 | XP_014979252.2 | 968 | 103 | ||
| XM_015123767.2 | XP_014979253.2 | 394 | 103 | |||
| XM_015123768.2 | XP_014979254.2 | 400 | 103 | |||
| XM_015123769.2 | XP_014979255.2 | 364 | 103 | |||
| XM_015123771.2 | XP_014979257.2 | 329 | 103 | |||
| XM_015123772.2 | XP_014979258.2 | 332 | 103 | |||
| XM_015123773.2 | XP_014979259.2 | 302 | 103 | |||
| XM_015123774.2 | XP_014979260.2 | 340 | 103 | |||
| GenBank mRNA | JV704285.1 | - | 324 | 103 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015123774.2 transcripts.
Set Membership: |
|

ZNF529
Genomic Map