Assay Details
| Gene Symbol: CENPBD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015126850.2 | XP_014982336.2 | 2216 | 109 | ||
| GenBank mRNA | JU350640.1 | - | 637 | 109 | ||
| JV727008.1 | - | 612 | 109 | |||
More Information
Set Membership: |
|

CENPBD1
Genomic Map