Assay Details
| Gene Symbol: VPS37B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001099814.4 | XP_001099814.1 | 1142 | 78 | ||
| XM_015153046.2 | XP_015008532.1 | 672 | 78 | |||
| XM_028830051.1 | XP_028685884.1 | 486 | 78 | |||
| GenBank mRNA | JU322755.1 | - | 295 | 78 | ||
More Information
Set Membership: |
|

VPS37B
Genomic Map