Assay Details
| Gene Symbol: APRT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001089867.4 | XP_001089867.2 | 256 | 86 | ||
| XM_015126795.2 | XP_014982281.2 | 256 | 86 | |||
| XM_028841000.1 | XP_028696833.1 | 256 | 86 | |||
| GenBank mRNA | JU332186.1 | - | 210 | 86 | ||
| JV567079.1 | - | 223 | 86 | |||
| JV639374.1 | - | 220 | 86 | |||
More Information
Set Membership: |
|

APRT
Genomic Map