Assay Details
| Gene Symbol: TSSK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001103323.3 | XP_001103323.1 | 866 | 92 | ||
| GenBank mRNA | JU494294.1 | - | 127 | 92 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TSSK3| Gene Symbol: TSSK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001103323.3 | XP_001103323.1 | 866 | 92 | ||
| GenBank mRNA | JU494294.1 | - | 127 | 92 | ||
Set Membership: |
|