Assay Details
| Gene Symbol: MDFIC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015134770.2 | XP_014990256.1 | 1207 | 67 | ||
| XM_015134771.2 | XP_014990257.1 | 1204 | 67 | |||
| GenBank mRNA | JU342528.1 | - | 417 | 67 | ||
| JU485260.1 | - | 636 | 67 | |||
More Information
Set Membership: |
|

MDFIC
Genomic Map