Assay Details
| Gene Symbol: ZDHHC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015126726.2 | XP_014982212.1 | 857 | 81 | ||
| XM_015126727.2 | XP_014982213.1 | 559 | 81 | |||
| GenBank mRNA | JU319875.1 | - | 464 | 81 | ||
| JU472905.1 | - | 457 | 81 | |||
| JV044574.1 | - | 483 | 81 | |||
| JV601787.1 | - | 653 | 81 | |||
More Information
Set Membership: |
|

ZDHHC7
Genomic Map