Assay Details
| Gene Symbol: CNST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015127739.2 | XP_014983225.1 | 1077 | 76 | ||
| XM_015127742.2 | XP_014983228.1 | 1040 | 76 | |||
| XM_015127747.2 | XP_014983233.1 | 861 | 76 | |||
| XM_015127758.2 | XP_014983244.1 | 832 | 76 | |||
| XM_028849102.1 | XP_028704935.1 | 1077 | 76 | |||
| XM_028849111.1 | XP_028704944.1 | 222 | 76 | |||
| XM_028849115.1 | XP_028704948.1 | 137 | 76 | |||
| GenBank mRNA | JU333191.1 | - | 753 | 76 | ||
| JU474505.1 | - | 1075 | 76 | |||
| JV046317.1 | - | 905 | 76 | |||
| JV643907.1 | - | 911 | 76 | |||
More Information
Set Membership: |
|

CNST
Genomic Map