Assay Details
| Gene Symbol: LRFN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028833182.1 | XP_028689015.1 | 2718 | 78 | ||
| GenBank mRNA | JV481436.1 | - | 319 | 78 | ||
| JV557449.1 | - | 330 | 78 | |||
More Information
Set Membership: |
|

LRFN4
Genomic Map