Assay Details
| Gene Symbol: CHRM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015134982.2 | XP_014990468.1 | 580 | 88 | ||
| XM_015134983.2 | XP_014990469.1 | 658 | 88 | |||
| XM_015134984.2 | XP_014990470.1 | 790 | 88 | |||
| XM_015134985.2 | XP_014990471.1 | 868 | 88 | |||
| GenBank mRNA | JU331122.1 | - | 117 | 88 | ||
More Information
Set Membership: |
|

CHRM2
Genomic Map