Assay Details
| Gene Symbol: ALG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015131478.2 | XP_014986964.2 | 980 | 67 | ||
| XM_015131479.2 | XP_014986965.2 | 943 | 67 | |||
| XM_015131480.2 | XP_014986966.2 | 1062 | 67 | |||
| XM_028843897.1 | XP_028699730.1 | 929 | 67 | |||
| GenBank mRNA | JU322676.1 | - | 947 | 67 | ||
| JU473909.1 | - | 947 | 67 | |||
| JV049357.1 | - | 804 | 67 | |||
| JV635901.1 | - | 1019 | 67 | |||
More Information
Set Membership: |
|

ALG3
Genomic Map