Assay Details
| Gene Symbol: SETMAR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001099426.4 | XP_001099426.2 | 1086 | 108 | ||
| XM_028844387.1 | XP_028700220.1 | 1168 | 108 | |||
| GenBank mRNA | JV045104.1 | - | 1041 | 108 | ||
More Information
Set Membership: |
|

SETMAR
Genomic Map