Assay Details
| Gene Symbol: HCN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002804859.3 | XP_002804905.2 | 1774 | 81 | ||
| GenBank mRNA | JV602793.1 | - | 231 | 81 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HCN4| Gene Symbol: HCN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002804859.3 | XP_002804905.2 | 1774 | 81 | ||
| GenBank mRNA | JV602793.1 | - | 231 | 81 | ||
Set Membership: |
|