Assay Details
| Gene Symbol: YDJC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001114556.4 | XP_001114556.3 | 421 | 66 | ||
| XM_015149793.2 | XP_015005279.2 | 424 | 66 | |||
| XM_028827942.1 | XP_028683775.1 | 419 | 66 | |||
| XM_028827943.1 | XP_028683776.1 | 419 | 66 | |||
| GenBank mRNA | JU322616.1 | - | 340 | 66 | ||
| JV707790.1 | - | 355 | 66 | |||
More Information
Set Membership: |
|

YDJC
Genomic Map