Assay Details
| Gene Symbol: ZNF543 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002801415.3 | XP_002801461.2 | 2100 | 65 | ||
| GenBank mRNA | JU325428.1 | - | 320 | 65 | ||
| JV706756.1 | - | 388 | 65 | |||
More Information
Set Membership: |
|

ZNF543
Genomic Map