Assay Details
| Gene Symbol: ZNF532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001089453.4 | XP_001089453.2 | 4108 | 90 | ||
| XM_002800954.3 | XP_002801000.1 | 4186 | 90 | |||
| XM_015122100.2 | XP_014977586.1 | 5010 | 90 | |||
| XM_015122101.2 | XP_014977587.1 | 4606 | 90 | |||
| XM_015122102.2 | XP_014977588.1 | 4607 | 90 | |||
| XM_015122104.2 | XP_014977590.1 | 5000 | 90 | |||
| XM_015122106.2 | XP_014977592.1 | 4993 | 90 | |||
| XM_015122107.2 | XP_014977593.1 | 4840 | 90 | |||
| XM_028838115.1 | XP_028693948.1 | 3803 | 90 | |||
| XM_028838116.1 | XP_028693949.1 | 3725 | 90 | |||
| XM_028838117.1 | XP_028693950.1 | 4498 | 90 | |||
| XM_028838118.1 | XP_028693951.1 | 3796 | 90 | |||
| XM_028838119.1 | XP_028693952.1 | 4597 | 90 | |||
| XM_028838120.1 | XP_028693953.1 | 3931 | 90 | |||
| XM_028838121.1 | XP_028693954.1 | 4830 | 90 | |||
| XM_028838122.1 | XP_028693955.1 | 4427 | 90 | |||
| XM_028838123.1 | XP_028693956.1 | 4248 | 90 | |||
| XM_028838124.1 | XP_028693957.1 | 3845 | 90 | |||
| GenBank mRNA | JU321521.1 | - | 3614 | 90 | ||
| JU335263.1 | - | 3596 | 90 | |||
| JV044437.1 | - | 3595 | 90 | |||
| JV047524.1 | - | 3577 | 90 | |||
| JV638087.1 | - | 3650 | 90 | |||
More Information
Set Membership: |
|

ZNF532
Genomic Map