Assay Details
| Gene Symbol: CCDC71L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001090799.4 | XP_001090799.2 | 1788 | 71 | ||
| GenBank mRNA | JU481588.1 | - | 1313 | 71 | ||
| JU481589.1 | - | 1313 | 71 | |||
More Information
Set Membership: |
|

CCDC71L
Genomic Map