Assay Details
| Gene Symbol: NNAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001091703.4 | XP_001091703.1 | 1088 | 77 | ||
| GenBank mRNA | JU320860.1 | - | 170 | 77 | ||
| JV546129.1 | - | 276 | 77 | |||
| JV652484.1 | - | 74 | 77 | |||
More Information
Set Membership: |
|

NNAT
Genomic Map