Assay Details
| Gene Symbol: ZNF503 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001095797.4 | XP_001095797.2 | 2916 | 93 | ||
| GenBank mRNA | JV631319.1 | - | 1110 | 93 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF503| Gene Symbol: ZNF503 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001095797.4 | XP_001095797.2 | 2916 | 93 | ||
| GenBank mRNA | JV631319.1 | - | 1110 | 93 | ||
Set Membership: |
|