Assay Details
| Gene Symbol: SLC36A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001110184.4 | XP_001110184.1 | 565 | 64 | ||
| XM_015141259.2 | XP_014996745.1 | 522 | 64 | |||
| XM_015141260.2 | XP_014996746.1 | 565 | 64 | |||
| XM_015141261.2 | XP_014996747.1 | 351 | 64 | |||
| XM_028849850.1 | XP_028705683.1 | 354 | 64 | |||
| XM_028849851.1 | XP_028705684.1 | 202 | 64 | |||
| XM_028849852.1 | XP_028705685.1 | 565 | 64 | |||
| XM_028849853.1 | XP_028705686.1 | 243 | 64 | |||
| GenBank mRNA | JV574199.1 | - | 534 | 64 | ||
| JV608649.1 | - | 536 | 64 | |||
More Information
Set Membership: |
|

SLC36A1
Genomic Map