Assay Details
| Gene Symbol: SIRPB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015149327.2 | XP_015004813.1 | 2141 | 69 | ||
| GenBank mRNA | JV712736.1 | - | 1020 | 69 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SIRPB2| Gene Symbol: SIRPB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015149327.2 | XP_015004813.1 | 2141 | 69 | ||
| GenBank mRNA | JV712736.1 | - | 1020 | 69 | ||
Set Membership: |
|