Assay Details
| Gene Symbol: SLC38A8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015126703.2 | XP_014982189.2 | 1029 | 57 | ||
| XM_028840787.1 | XP_028696620.1 | 1881 | 57 | |||
| GenBank mRNA | JU480797.1 | - | 911 | 57 | ||
| JV707530.1 | - | 679 | 57 | |||
More Information
Set Membership: |
|

SLC38A8
Genomic Map