Assay Details
| Gene Symbol: KRT24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015119657.2 | XP_014975143.1 | 1496 | 93 | ||
| GenBank mRNA | JU327810.1 | - | 38 | 93 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRT24| Gene Symbol: KRT24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015119657.2 | XP_014975143.1 | 1496 | 93 | ||
| GenBank mRNA | JU327810.1 | - | 38 | 93 | ||
Set Membership: |
|