Assay Details
| Gene Symbol: PURG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015145122.2 | XP_015000608.1 | 1604 | 104 | ||
| GenBank mRNA | JU342511.1 | - | 226 | 104 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PURG| Gene Symbol: PURG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015145122.2 | XP_015000608.1 | 1604 | 104 | ||
| GenBank mRNA | JU342511.1 | - | 226 | 104 | ||
Set Membership: |
|