Assay Details
| Gene Symbol: KLK8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015124679.2 | XP_014980165.2 | 421 | 87 | ||
| XM_028838918.1 | XP_028694751.1 | 374 | 87 | |||
| XM_028838919.1 | XP_028694752.1 | 366 | 87 | |||
| GenBank mRNA | EF407409.1 | - | 368 | 87 | ||
| EU350537.1 | - | 505 | 87 | |||
| JV641152.1 | - | 91 | 87 | |||
More Information
Set Membership: |
|

KLK8
Genomic Map