Assay Details
| Gene Symbol: YJEFN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015123396.2 | XP_014978882.1 | 839 | 61 | ||
| XM_015123397.2 | XP_014978883.1 | 728 | 61 | |||
| XM_028840088.1 | XP_028695921.1 | 806 | 61 | |||
| GenBank mRNA | JU321397.1 | - | 462 | 61 | ||
| JV602216.1 | - | 463 | 61 | |||
More Information
Set Membership: |
|

YJEFN3
Genomic Map