Assay Details
| Gene Symbol: CMTM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002804989.3 | XP_002805035.1 | 802 | 100 | ||
| XM_015143410.2 | XP_014998896.1 | 895 | 100 | |||
| XM_015143412.2 | XP_014998898.1 | 743 | 100 | |||
| XM_015143413.2 | XP_014998899.1 | 650 | 100 | |||
| GenBank mRNA | JU323396.1 | - | 904 | 100 | ||
| JU323475.1 | - | 811 | 100 | |||
More Information
Set Membership: |
|

CMTM5
Genomic Map