Assay Details
| Gene Symbol: CCZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015133273.2 | XP_014988759.1 | 1404 | 123 | ||
| XM_028845138.1 | XP_028700971.1 | 839 | 123 | |||
| GenBank mRNA | JU334942.1 | - | 863 | 123 | ||
| JU475946.1 | - | 872 | 123 | |||
| JU475947.1 | - | 916 | 123 | |||
| JV584224.1 | - | 834 | 123 | |||
| JV584225.1 | - | 820 | 123 | |||
| Gene Symbol: LOC106995287 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028845111.1 | XP_028700944.1 | 1191 | 123 | ||
More Information
Set Membership: |
|

CCZ1
Genomic Map