Assay Details
| Gene Symbol: NUDT6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001103787.4 | XP_001103787.2 | 607 | 92 | ||
| XM_015139179.2 | XP_014994665.1 | 607 | 92 | |||
| GenBank mRNA | JV574739.1 | - | 687 | 92 | ||
More Information
Set Membership: |
|

NUDT6
Genomic Map