Assay Details
| Gene Symbol: FAM83G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015118773.2 | XP_014974259.1 | 1018 | 79 | ||
| GenBank mRNA | JV068689.1 | - | 145 | 79 | ||
| JV658854.1 | - | 28 | 79 | |||
More Information
Set Membership: |
|

FAM83G
Genomic Map