Assay Details
| Gene Symbol: MAFB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001101624.4 | XP_001101624.3 | 1007 | 111 | ||
| GenBank mRNA | JU320635.1 | - | 621 | 111 | ||
| JU478089.1 | - | 618 | 111 | |||
More Information
Set Membership: |
|

MAFB
Genomic Map