Assay Details
| Gene Symbol: OCM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015134346.2 | XP_014989832.2 | 424 | 74 | ||
| GenBank mRNA | JV652651.1 | - | 151 | 74 | ||
| Gene Symbol: OCM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028845177.1 | XP_028701010.1 | 322 | 74 | ||
More Information
Set Membership: |
|

OCM
Genomic Map