Assay Details
| Gene Symbol: FFAR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015147708.2 | XP_015003194.1 | 717 | 123 | ||
| GenBank mRNA | JV615189.1 | - | 221 | 123 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FFAR4| Gene Symbol: FFAR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015147708.2 | XP_015003194.1 | 717 | 123 | ||
| GenBank mRNA | JV615189.1 | - | 221 | 123 | ||
Set Membership: |
|