Assay Details
| Gene Symbol: SETBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002808161.3 | XP_002808207.3 | 1190 | 75 | ||
| XM_028837859.1 | XP_028693692.1 | 850 | 75 | |||
| XM_028837860.1 | XP_028693693.1 | 3536 | 75 | |||
| GenBank mRNA | JV711652.1 | - | 778 | 75 | ||
More Information
Set Membership: |
|

SETBP1
Genomic Map