Assay Details
| Gene Symbol: ZC3H12D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001087043.4 | XP_001087043.2 | 628 | 81 | ||
| XM_015137149.2 | XP_014992635.1 | 596 | 81 | |||
| GenBank mRNA | JU485694.1 | - | 545 | 81 | ||
More Information
Set Membership: |
|

ZC3H12D
Genomic Map