Assay Details
| Gene Symbol: TRIM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015134414.2 | XP_014989900.2 | 909 | 58 | ||
| XM_015134415.2 | XP_014989901.2 | 909 | 58 | |||
| GenBank mRNA | JU326316.1 | - | 526 | 58 | ||
| JU471814.1 | - | 887 | 58 | |||
| JV044938.1 | - | 902 | 58 | |||
| JV635738.1 | - | 921 | 58 | |||
More Information
Set Membership: |
|

TRIM4
Genomic Map