Assay Details
| Gene Symbol: ZNF841 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015124743.2 | XP_014980229.2 | 531 | 130 | ||
| XM_028839356.1 | XP_028695189.1 | 239 | 130 | |||
| GenBank mRNA | JV648896.1 | - | 97 | 130 | ||
| JV712666.1 | - | 95 | 130 | |||
More Information
Set Membership: |
|

ZNF841
Genomic Map