Assay Details
| Gene Symbol: LOC703985 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015117567.2 | XP_014973053.2 | 544 | 106 | ||
| XM_015117568.2 | XP_014973054.2 | 333 | 106 | |||
| XM_015117570.2 | XP_014973056.2 | 419 | 106 | |||
| XM_015117571.2 | XP_014973057.2 | 270 | 106 | |||
| XM_015117573.2 | XP_014973059.2 | 242 | 106 | |||
| XM_028835110.1 | XP_028690943.1 | 414 | 106 | |||
| XM_028835111.1 | XP_028690944.1 | 265 | 106 | |||
| GenBank mRNA | JU324953.1 | - | 119 | 106 | ||
| JV045092.1 | - | 152 | 106 | |||
| JV637943.1 | - | 153 | 106 | |||
More Information
Set Membership: |
|

LOC703985
Genomic Map