Assay Details
| Gene Symbol: COQ4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015116724.2 | XP_014972210.2 | 715 | 94 | ||
| GenBank mRNA | JU344302.1 | - | 240 | 94 | ||
| JU475827.1 | - | 647 | 94 | |||
| JV047252.1 | - | 663 | 94 | |||
| JV636195.1 | - | 676 | 94 | |||
More Information
Set Membership: |
|

COQ4
Genomic Map