Assay Details
| Gene Symbol: WIPF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028845903.1 | XP_028701736.1 | 1378 | 92 | ||
| XM_028845904.1 | XP_028701737.1 | 1308 | 92 | |||
| GenBank mRNA | JV590751.1 | - | 216 | 92 | ||
| JV721164.1 | - | 307 | 92 | |||
More Information
Set Membership: |
|

WIPF3
Genomic Map