Assay Details
| Gene Symbol: ZNF616 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002801385.3 | XP_002801431.2 | 1191 | 96 | ||
| GenBank mRNA | JV726359.1 | - | 517 | 96 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF616| Gene Symbol: ZNF616 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002801385.3 | XP_002801431.2 | 1191 | 96 | ||
| GenBank mRNA | JV726359.1 | - | 517 | 96 | ||
Set Membership: |
|