Assay Details
| Gene Symbol: CHST12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028845502.1 | XP_028701335.1 | 496 | 126 | ||
| XM_028845503.1 | XP_028701336.1 | 608 | 126 | |||
| GenBank mRNA | JU476293.1 | - | 418 | 126 | ||
| JV621446.1 | - | 418 | 126 | |||
| JV639054.1 | - | 418 | 126 | |||
More Information
Set Membership: |
|

CHST12
Genomic Map