Assay Details
Gene Symbol: TTC30B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015110423.2 | XP_014965909.2 | 2501 | 106 | ||
GenBank mRNA | JU326430.1 | - | 1924 | 106 | ||
JU339336.1 | - | 2270 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
