Assay Details
| Gene Symbol: ZFP69B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001083211.4 | XP_001083211.2 | 1121 | 129 | ||
| XM_015136493.2 | XP_014991979.1 | 1035 | 129 | |||
| GenBank mRNA | JU329247.1 | - | 320 | 129 | ||
| JV721496.1 | - | 452 | 129 | |||
More Information
Set Membership: |
|

ZFP69B
Genomic Map