Assay Details
| Gene Symbol: ACKR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001117200.4 | XP_001117200.2 | 944 | 132 | ||
| GenBank mRNA | JU475021.1 | - | 824 | 132 | ||
| JV711176.1 | - | 958 | 132 | |||
More Information
Set Membership: |
|

ACKR1
Genomic Map