Assay Details
| Gene Symbol: C19H19orf53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015123095.2 | XP_014978581.2 | 353 | 117 | ||
| GenBank mRNA | JU332111.1 | - | 324 | 117 | ||
| JV489615.1 | - | 311 | 118 | |||
| JV529075.1 | - | 329 | 118 | |||
| JV667306.1 | - | 321 | 117 | |||
| JV704671.1 | - | 329 | 117 | |||
More Information
Set Membership: |
|

C19H19orf53
Genomic Map