Assay Details
| Gene Symbol: LOC100425237 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002804975.3 | XP_002805021.2 | 344 | 95 | ||
| GenBank mRNA | JU470678.1 | - | 311 | 95 | ||
More Information
Set Membership: |
|

LOC100425237
Genomic Map